missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon (IV) oxide, 99.5%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 13024A9

 View more versions of this product

Catalog No. AA13024A9



Specifications

Specifications

Silicon(IV) oxide
14808-60-7
O2Si
aerosil
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
2.2-2.6g/mL
2230°C
1710°C
98.5-99% microcrystalline silicon in the form of quartz
99.5%
MFCD00011232
Insoluble in water and acids. Soluble in HF. Very slightly soluble in alkalies
O=[Si]=O
60.083
CHEBI:30563
<10 Micron APS Powder
Odorless
Brown to White
10kg

Chemical Identifiers

14808-60-7
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
aerosil
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H350

EINECSNumber : 238-878-4

RTECSNumber : VV7330000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents