Please login to your online account to display your discounted pricing

Silicon(IV) oxide, 40% in H2O, Colloidal dispersion, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Silicon(IV) oxide
Name Note 40% in H2O
CAS 7631-86-9
Physical Form Liquid
Molecular Formula SiO2
Formula Weight 60.09
MDL Number MFCD00011232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA12727A1

 
alfa aesar
12727A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA12727A7

 
alfa aesar
12727A7
5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.09
Quantity 1kg
Color Colorless
Physical Form Liquid
Chemical Name or Material Silicon(IV) oxide
Odor Odorless
Name Note 40% in H2O
Molecular Formula SiO2
MDL Number MFCD00011232
CAS 7631-86-9
Health Hazard 1 Flammable.
Health Hazard 2 Take precautionary measures against static discharges.
Density 1.3g/cm3 at 20°C
Safety and Handling

Flammable.

Take precautionary measures against static discharges.