Please login to your online account to display your discounted pricing

Silicon(IV) oxide, 40% in H2O, 0.02 micron particles in colloidal dispersion, Alfa Aesar™

S.A. 200 m2/g

$39.70 - $115.30

Chemical Identifiers

Chemical Name or Material Silicon(IV) oxide
Name Note 40% in H2O
CAS 7631-86-9
Physical Form Liquid
Molecular Formula SiO2
Formula Weight 60.09
MDL Number MFCD00011232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43110A1

 
alfa aesar
43110A1
1kg Each for $39.70

AA43110A7

 
alfa aesar
43110A7
5kg Each for $115.30
Description & Specifications

Specifications

Formula Weight 60.09
Quantity 1kg
Color Colorless
Density 1.30g/cm3 at 20°C
Chemical Name or Material Silicon(IV) oxide
EINECS Number 231-545-4
Odor Odorless
Name Note 40% in H2O
Physical Form Liquid
Molecular Formula SiO2
MDL Number MFCD00011232
CAS 7631-86-9