Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) nitride, MgO binder, sputtering target, 50.8mm (2.0in.) dia x 3.18mm (0.125in.) thick, 99.9% (metals basis)

Magnetron sputtering source

Chemical Identifiers

Chemical Name or Material Silicon(IV) nitride, MgO binder, Sputtering Target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 140.28
MDL Number MFCD00011230
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41088KS

 
alfa aesar™
41088KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.28
Chemical Name or Material Silicon(IV) nitride, MgO binder, Sputtering Target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011230
Quantity 1 each