missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) nitride, Electronic Grade, 99.85% (metals basis), 90+% alpha-phase

Manufacturer:  Alfa Aesar™ 4911318

 View more versions of this product

Catalog No. AA4911318



Specifications

Specifications

Silicon (IV) nitride
12033-89-5
99.85%
N4Si3
MFCD00011230
HQVNEWCFYHHQES-UHFFFAOYSA-N
140.283
140.28
3.2g/mL
Odorless
1900°C (sublimation)
≥90% alpha-phase
Electronic Grade
(metals basis)
Si3N4
14,8497
N12[Si]34N5[Si]16N3[Si]25N46
3084099
Powder
−325 Mesh
Gray
50g

Chemical Identifiers

12033-89-5
140.283
HQVNEWCFYHHQES-UHFFFAOYSA-N
N12[Si]34N5[Si]16N3[Si]25N46
N4Si3
MFCD00011230
3084099
Safety and Handling

Safety and Handling

RTECSNumber : VW0650000

TSCA : TSCA

SDS Documents