missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) nitride, amorphous, nanopowder, 98.5+%

Manufacturer:  Alfa Aesar™ 4561106

 View more versions of this product

Catalog No. AA4561106



Specifications

Specifications

Silicon (IV) nitride
12033-89-5
MFCD00011230
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
N12[Si]34N5[Si]16N3[Si]25N46
3084099
Nanopowder
3.2g/mL
15 to 30nm APS
Gray
5g
Amorphous
N4Si3
14,8497
HQVNEWCFYHHQES-UHFFFAOYSA-N
140.283
140.28
≥98.5%
Odorless
103 to 123m2/g
1900°C (sublimation)

Chemical Identifiers

12033-89-5
140.283
HQVNEWCFYHHQES-UHFFFAOYSA-N
3084099
N4Si3
MFCD00011230
silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
N12[Si]34N5[Si]16N3[Si]25N46
Safety and Handling

Safety and Handling

RTECSNumber : VW0650000

TSCA : TSCA

SDS Documents