missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon(IV) iodide, 99% (metals basis), Alfa Aesar™

Light, Air and Moisture Sensitive

Manufacturer:  Alfa Aesar 8730314

 View more versions of this product

Catalog No. AA8730314



Specifications

Specifications

Silicon(IV) iodide
99% (metals basis)
MFCD00054041
CFTHARXEQHJSEH-UHFFFAOYSA-N
tetraiodosilane
83498
Crystals, Packaged Under Argon
25g
13465-84-4
I4Si
silicon tetraiodide, silane, tetraiodo, silicon iv iodide, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
[Si](I)(I)(I)I
535.703
535.7
White

Chemical Identifiers

13465-84-4
535.703
CFTHARXEQHJSEH-UHFFFAOYSA-N
83498
[Si](I)(I)(I)I
I4Si
MFCD00054041
silicon tetraiodide, silane, tetraiodo, silicon iv iodide, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
tetraiodosilane
Safety and Handling

Safety and Handling

H300-H314-H318: Fatal if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 236-706-2

SDS Documents