missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) iodide, 99.999% (metals basis)

$225.78 - $2,403.28

Chemical Identifiers

CAS 13465-84-4
Molecular Formula I4Si
Molecular Weight (g/mol) 535.703
MDL Number MFCD00054041
InChI Key CFTHARXEQHJSEH-UHFFFAOYSA-N
Synonym acmc-1bu37, periodosilane, silane, tetraiodo, silicon iv iodide, silicon iv iodide trace metals basis 1g, silicon tetraiodide, silicon tetraiodide trace metals basis, tetraiodosilicon
PubChem CID 83498
IUPAC Name tetraiodosilane
SMILES [Si](I)(I)(I)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4414106 View Documents Alfa Aesar™
4414106
5g Each for $225.78
 
AA4414114 View Documents Alfa Aesar™
4414114
25g Each for $803.95
 
AA4414122 View Documents Alfa Aesar™
4414122
100g Each for $2,403.28
 
Description

Description

  • Packaged under Argon
Specifications

Chemical Identifiers

13465-84-4
535.703
CFTHARXEQHJSEH-UHFFFAOYSA-N
83498
[Si](I)(I)(I)I
I4Si
MFCD00054041
acmc-1bu37, periodosilane, silane, tetraiodo, silicon iv iodide, silicon iv iodide trace metals basis 1g, silicon tetraiodide, silicon tetraiodide trace metals basis, tetraiodosilicon
tetraiodosilane

Specifications

13465-84-4
SiI4
UN2923
Soluble in organic solvents. Insoluble in water.
[Si](I)(I)(I)I
535.703
535.7
99.999%
Air, light, and moisture sensitive
White
5g
(metals basis)
I4Si
MFCD00054041
acmc-1bu37, periodosilane, silane, tetraiodo, silicon iv iodide, silicon iv iodide trace metals basis 1g, silicon tetraiodide, silicon tetraiodide trace metals basis, tetraiodosilicon
CFTHARXEQHJSEH-UHFFFAOYSA-N
tetraiodosilane
83498
Lumps
4.198 g/mL
290°C
120°C
Silicon (IV) iodide
Safety and Handling

Safety and Handling

GHS H Statement
H300-H314-H318
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 236-706-2

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Nitrogen