missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon(IV) chloride, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 69182AE

 View more versions of this product

Catalog No. AA69182AE



Specifications

Specifications

Silicon(IV) chloride
99% (metals basis)
MFCD00011229
silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
[Si](Cl)(Cl)(Cl)Cl
169.885
169.9
Acrid
100mL
10026-04-7
Cl4Si
Colourless liquid
FDNAPBUWERUEDA-UHFFFAOYSA-N
tetrachlorosilane
24816
Liquid
Colorless

Chemical Identifiers

10026-04-7
169.885
FDNAPBUWERUEDA-UHFFFAOYSA-N
24816
[Si](Cl)(Cl)(Cl)Cl
Cl4Si
MFCD00011229
silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
tetrachlorosilane
Safety and Handling

Safety and Handling

H314-H319-H335: Causes severe skin burns and eye damage. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 233-054-0

SDS Documents