Learn More
Silicon(IV) chloride, 99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 069182AB
Description
Silicon(IV) chloride is used as an intermediate in the manufacture of polysilicon. It is also used in the semiconductors industry, photovoltaic cells and in the production of optical fibers. Its derivatives like trichlorosilane and silane are used in the Siemens process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
57.6°C | |
°C to 70°C | |
Colorless | |
10 mL | |
Cl4Si | |
silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4 | |
FDNAPBUWERUEDA-UHFFFAOYSA-N | |
tetrachlorosilane | |
24816 | |
Acrid | |
Colourless liquid |
Silicon(IV) chloride | |
10026-04-7 | |
Liquid | |
99% (metals basis) | |
MFCD00011229 | |
Miscible with benzene,toluene,chloroform,petroleum ether,carbon tetrachloride,ether and hydrochloric acid. | |
Cl[Si](Cl)(Cl)Cl | |
169.89 | |
169.9 | |
Moisture sensitive | |
1.483 |
Chemical Identifiers
10026-04-7 | |
169.89 | |
FDNAPBUWERUEDA-UHFFFAOYSA-N | |
24816 | |
Cl[Si](Cl)(Cl)Cl |
Cl4Si | |
MFCD00011229 | |
silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4 | |
tetrachlorosilane |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H331-H314-H335
DOTInformation : DOT Class: 8, Packaging Group: II
EINECSNumber : 233-054-0
RTECSNumber : VW0525000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only