Learn More
Silicon(IV) chloride, 99.998% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 040148AE
Description
Silicon(IV) chloride is used as semiconductor grade.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
57.6°C | |
-70°C | |
Colorless | |
100 mL | |
Cl4Si | |
MFCD00011229 | |
14,8500 | |
Reacts with water. Miscible with benzene,ether,chloroform. | |
Cl[Si](Cl)(Cl)Cl | |
169.89 | |
169.9 | |
Acrid | |
1.483 g/mL |
Silicon(IV) chloride | |
10026-04-7 | |
Liquid | |
(metals basis) | |
SiCl4 | |
UN1818 | |
silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4 | |
FDNAPBUWERUEDA-UHFFFAOYSA-N | |
tetrachlorosilane | |
24816 | |
99.998% | |
Moisture sensitive |
Chemical Identifiers
10026-04-7 | |
169.89 | |
FDNAPBUWERUEDA-UHFFFAOYSA-N | |
24816 | |
Cl[Si](Cl)(Cl)Cl |
Cl4Si | |
MFCD00011229 | |
silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4 | |
tetrachlorosilane |
Safety and Handling
GHS H Statement
H314-H335-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H331-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SILICON TETRACHLORIDE
EINECSNumber : 233-054-0
RTECSNumber : VW0525000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only