missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) bromide

Manufacturer:  Alfa Aesar™ 1402918

 View more versions of this product

Catalog No. AA1402918



Specifications

Specifications

Silicon(IV) bromide
Br4Si
UN3264
silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, silicon iv bromide, acmc-20ajqj, silicon tetrabromide
[Si](Br)(Br)(Br)Br
347.701
347.72
2.800g/mL
Moisture Sensitive
Colorless to Yellow
50g
7789-66-4
MFCD00049530
14,8499
AIFMYMZGQVTROK-UHFFFAOYSA-N
tetrabromosilane
82247
Liquid
Stench
154°C
5°C

Chemical Identifiers

7789-66-4
347.701
AIFMYMZGQVTROK-UHFFFAOYSA-N
82247
[Si](Br)(Br)(Br)Br
Br4Si
MFCD00049530
silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, silicon iv bromide, acmc-20ajqj, silicon tetrabromide
tetrabromosilane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents