Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) 2-ethylhexanoate, nominally 75% in ethanol

Chemical Identifiers

Chemical Name or Material Silicon (IV) 2-ethylhexanoate
CAS 67939-81-5
Molecular Formula C32H60O8Si
Formula Weight 600.91
MDL Number MFCD00269842
UN Number UN1170
Synonym 2-Ethylhexanoic acid tetraanhydride with silicic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4463530

  SDS
alfa aesar™
4463530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 600.91
Chemical Name or Material Silicon (IV) 2-ethylhexanoate
Concentration Nominally 75% in ethanol
TSCA TSCA
Sensitivity Moisture Sensitive
Physical Form Liquid
Molecular Formula C32H60O8Si
MDL Number MFCD00269842
CAS 67939-81-5
Color Yellow
UN Number UN1170
Synonym 2-Ethylhexanoic acid tetraanhydride with silicic acid
Health Hazard 1 Danger
Quantity 250g
Health Hazard 2 GHS H Statement
H225-H361
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
P210-P280h
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/clothing.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Safety and Handling

Danger

GHS P Statement
P210-P280h
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/clothing.

GHS H Statement
H225-H361
Highly flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.