Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(II) oxide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.9% (metals basis)

$1,378.69 - $1,378.69

Chemical Identifiers

Chemical Name or Material Silicon(II) oxide sputtering target
CAS 10097-28-6
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 44.09
MDL Number MFCD00151536
Synonym Oxosilylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41093KS

  SDS
alfa aesar™
41093KS
1 each Each for $1,378.69
Description & Specifications

Specifications

Formula Weight 44.09
Chemical Name or Material Silicon(II) oxide sputtering target
TSCA TSCA
Odor Odorless
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Physical Form Target
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00151536
CAS 10097-28-6
Synonym Oxosilylene
Boiling Point 1880°C
Melting Point >1702°C
Quantity 1 each
Density 2.13g/mL