Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(II) oxide, Optical Grade, 99.8% (metals basis)

Chemical Identifiers

Chemical Name or Material Silicon(II) oxide
CAS 10097-28-6
Purity Grade Optical Grade
Linear Formula SiO
Formula Weight 44.09g/mol
MDL Number MFCD00151536
Merck Index 14,8496
Synonym Oxosilylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2272918

 
alfa aesar™
22729-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2272930

 
alfa aesar™
22729-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 2.13g/mL
Mesh Size −40+80 Mesh
Odor Odorless
Boiling Point 1880°C
Color Black
Melting Point >1702°C
Quantity 50g
Chemical Name or Material Silicon(II) oxide
CAS 10097-28-6
Purity Grade Optical Grade
Assay 99.8%
Assay Percent Notes (metals basis)
Linear Formula SiO
Formula Weight 44.09g/mol
MDL Number MFCD00151536
Merck Index 14,8496
Synonym Oxosilylene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.