missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(II) oxide, Optical Grade, 99.7% (metals basis)

Manufacturer:  Alfa Aesar™ 8982918

 View more versions of this product

Catalog No. AA8982918



Specifications

Specifications

Silicon (II) oxide
Optical Grade
(metals basis)
SiO
14,8496
LIVNPJMFVYWSIS-UHFFFAOYSA-N
oxoniumylidynesilanide
66241
44.09
−10 Mesh
1880°C
>1702°C
10097-28-6
99.7%
OSi
MFCD00151536
Oxosilylene
[O+]#[Si-]
44.084
CHEBI:30588
2.13g/mL
Odorless
Black
50g

Chemical Identifiers

10097-28-6
44.084
LIVNPJMFVYWSIS-UHFFFAOYSA-N
66241
oxoniumylidynesilanide
OSi
MFCD00151536
Oxosilylene
CHEBI:30588
[O+]#[Si-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents