missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon ICP Standard, 1mL = 10mg Si (10,000ppm Si), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPSI10KW500

 View more versions of this product

Catalog No. PSI10KW-500



Specifications

Specifications

Silicon ICP Standard
92.0,5.84,0.09
F6H8N2Si
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
178.153
Liquid
10,000ppm Si
Colorless
16919-19-0,7664-39-3,7732-18-5
96.0,6.08,0.1
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)
28145
Poly natural
1 mL = 10 mg Si
500mL

Chemical Identifiers

16919-19-0
178.153
28145
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
F6H8N2Si
ITHIMUMYFVCXSL-UHFFFAOYSA-P
diazanium;hexafluorosilicon(2-)
Documents