missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicon ICP Standard, 10,000 ppm Si in H2O/tr HF, Ricca Chemical
$447.97 - $1008.69
Chemical Identifiers
CAS | 16919-19-0 |
---|---|
Molecular Formula | F6H8N2Si |
Molecular Weight (g/mol) | 178.153 |
InChI Key | ITHIMUMYFVCXSL-UHFFFAOYSA-P |
PubChem CID | 28145 |
IUPAC Name | diazanium;hexafluorosilicon(2-) |
SMILES | [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
PSI10KW-100
|
Ricca Chemical Company
PSI10KW100 |
100 mL | Natural LDPE Bottle |
Each for $447.97
|
|
||||
PSI10KW-500
|
Ricca Chemical Company
PSI10KW500 |
500 mL | Natural Poly Bottle |
Each for $1,008.69
|
|
||||
Chemical Identifiers
16919-19-0 | |
178.153 | |
28145 | |
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F |
F6H8N2Si | |
ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
diazanium;hexafluorosilicon(2-) |
Specifications
16919-19-0,7664-39-3,7732-18-5 | |
100 mL | |
96.0,6.08,0.1 | |
10,000ppm Si | |
ITHIMUMYFVCXSL-UHFFFAOYSA-P | |
diazanium;hexafluorosilicon(2-) | |
28145 | |
Liquid |
Natural LDPE Bottle | |
92.0,5.84,0.09 | |
Colorless | |
F6H8N2Si | |
[NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F | |
178.153 | |
1 mL = 10 mg Si | |
Silicon ICP Standard |