missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon granular, virgin poly fines, 99.999% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03854214

 View more versions of this product

Catalog No. AA3854214


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silicon granular
7440-21-3
Si
25 g
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
99.999%
∽1/4 in. & down
Virgin poly fines
(metals basis)
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
[Si]
28.09
CHEBI:27573
Odorless
Chunks to Powder

Chemical Identifiers

7440-21-3
28.09
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
CHEBI:27573
[Si]
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 231-130-8

RTECSNumber : VW0400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only