missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon dioxide, 99.998%, (trace metal basis), Thermo Scientific Chemicals

For ultra trace analysis

Supplier:  Thermo Scientific Chemicals 437151000

 View more versions of this product

Catalog No. AC437151000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

Silicon dioxide
14808-60-7
Solution
44.5 to 47.9% (Si)
SiO2
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
60.08
Trace Metal Basis
1610.0°C
Colorless to Yellow
100 g
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
Solubility in water: insoluble
O=[Si]=O
60.08
CHEBI:30563
99.998%
Glass bottle

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
silanedione
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer by inhalation.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 238-878-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only