Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon carbide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis excluding B)

Chemical Identifiers

Chemical Name or Material Silicon carbide sputtering target
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 40.1
MDL Number MFCD00049531
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41086KS

 
alfa aesar™
41086KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 40.1
Chemical Name or Material Silicon carbide sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.5%
Assay Percent Notes (metals basis excluding B)
MDL Number MFCD00049531
Quantity 1 each