Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon carbide sputtering target, 25.4mm (1.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis excluding B)

Chemical Identifiers

Chemical Name or Material Silicon carbide sputtering target
Name Note 25.4mm (1.0 in.) dia. x 3.18mm (0.125 in.) thick
Molecular Formula SiC
Formula Weight 40.1
MDL Number MFCD00049531
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA44277KS

  SDS
alfa aesar™
44277KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 40.1
Assay Percent Notes (metals basis excluding B)
MDL Number MFCD00049531
Quantity 1 each
Chemical Name or Material Silicon carbide sputtering target
Molecular Formula SiC
Name Note 25.4mm (1.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.5%
TSCA TSCA