missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon carbide, beta-phase, nanopowder, 95% min

Manufacturer:  Alfa Aesar™ 4388406

 View more versions of this product

Catalog No. AA4388406



Specifications

Specifications

Silicon carbide
409-21-2
MFCD00049531
betarundum
HBMJWWWQQXIZIP-UHFFFAOYSA-N
methanidylidynesilanylium
9863
40.1
≥95%
<30nm APS
Black to Gray
5g
beta-phase
CSi
14,8492
Insoluble in water, alcohol; Soluble in fused alkali
[C-]#[Si+]
40.096
CHEBI:29390
Nanopowder
Odorless
>80m2/g
2700°C

Chemical Identifiers

409-21-2
40.096
HBMJWWWQQXIZIP-UHFFFAOYSA-N
9863
methanidylidynesilanylium
CSi
MFCD00049531
betarundum
CHEBI:29390
[C-]#[Si+]
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VW0450000

TSCA : TSCA

Documents