Learn More
Silicon carbide, beta-phase, nanopowder, Thermo Scientific Chemicals
S.A. 70 to 90 m2/g
Supplier: Thermo Scientific Chemicals 04464614
Description
Silicon carbide can be used form very hard ceramics that are widely used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide as light-emitting diodes (LEDs) and detectors in early radios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silicon carbide | |
beta-phase | |
409-21-2 | |
25 g | |
MFCD00049531 | |
Insoluble in water | |
[C-]#[Si+] | |
40.10 | |
CHEBI:29390 | |
Odorless |
2700°C | |
Black to Gray | |
Powder | |
CSi | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
methanidylidynesilylium | |
9863 | |
40.1 |
Chemical Identifiers
409-21-2 | |
40.10 | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
9863 | |
[C-]#[Si+] |
CSi | |
MFCD00049531 | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
CHEBI:29390 |
Safety and Handling
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H350
EINECSNumber : 206-991-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only