missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon carbide, alpha-phase, 99.8% (metals basis), Thermo Scientific Chemicals

Surface Area 9-11 m2/g

Supplier:  Thermo Scientific Chemicals 04015522

 View more versions of this product

Catalog No. AA4015522



Description

Description

Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silicon carbide
alpha-phase
409-21-2
100 g
CSi
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
HBMJWWWQQXIZIP-UHFFFAOYSA-N
methanidylidynesilylium
9863
40.1
2700°C
Black to Gray
Powder
99.8% (metals basis)
MFCD00049531
Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron.
[C-]#[Si+]
40.10
CHEBI:29390
Odorless

Chemical Identifiers

409-21-2
40.10
HBMJWWWQQXIZIP-UHFFFAOYSA-N
9863
[C-]#[Si+]
CSi
MFCD00049531
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
CHEBI:29390
Safety and Handling

Safety and Handling

H351-H315-H319-H335: Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 206-991-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only