Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon carbide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Silicon carbide
CAS 409-21-2
Formula Weight 40.1
MDL Number MFCD00049531
Merck Index 14,8492
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA9793836

  SDS
alfa aesar™
9793836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 40.1
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 2700°C
MDL Number MFCD00049531
RTECS Number VW0450000
Merck Index 14,8492
Particle Size ≈7 Micron
Odor Odorless
Chemical Name or Material Silicon carbide
CAS 409-21-2
Density 3.16g/mL
TSCA TSCA
Color Green
pH 3 to 7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay Percent Notes (metals basis)
Assay 99.5%
Physical Form Powder
Quantity 500g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.