Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon carbide, 600 grinding compound, 2oz (57g)

Grinding compound for polishing IR crystal windows

Chemical Identifiers

Chemical Name or Material Silicon carbide
CAS 409-21-2
Name Note 600 grinding compound
Formula Weight 40.1
MDL Number MFCD00049531
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39801LA

  SDS
alfa aesar™
39801LA
57g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 40.1
TSCA TSCA
Name Note 600 grinding compound
CAS 409-21-2
MDL Number MFCD00049531
Quantity 57g
Health Hazard 2 GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Weight 2oz (57g)
Chemical Name or Material Silicon carbide
Health Hazard 1 Warning
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.