Learn More
Silicon carbide, 400 grinding compound, 2oz (57g), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 039800LA
Description
Grinding compound for polishing IR crystal windows
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silicon carbide | |
400 grinding compound | |
Black to Gray | |
57 g | |
MFCD00049531 | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
methanidylidynesilylium | |
9863 | |
40.1 | |
3.16 g/mL |
2700°C | |
409-21-2 | |
Powder | |
CSi | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
[C-]#[Si+] | |
40.10 | |
CHEBI:29390 | |
Odorless |
Chemical Identifiers
409-21-2 | |
40.10 | |
HBMJWWWQQXIZIP-UHFFFAOYSA-N | |
9863 | |
methanidylidynesilylium |
CSi | |
MFCD00049531 | |
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide | |
CHEBI:29390 | |
[C-]#[Si+] |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 206-991-8
RTECSNumber : VW0450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only