missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon Carbide, 325 Mesh, 97.5%, Spectrum™

$251.63 - $7,477.29

Chemical Identifiers

CAS 409-21-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613108
View Documents
Spectrum Chemical Mfg Cor
S103325KG
2.5 kg Poly Pail
Each for $815.74
 
18613109
View Documents
Spectrum Chemical Mfg Cor
S103345KGBL
45 kg Fiber Drum
Each for $7,477.29
 
18613110
View Documents
Spectrum Chemical Mfg Cor
S1033500GM
500 g Poly Bottle
Each for $251.63
 
Description

Description

Specifications

Chemical Identifiers

409-21-2

Specifications

409-21-2
0.975
Poly Pail
1
Reagent
2.5 kg
SDS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.