Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon, AAS standard solution, Specpure™, Si 1000μg/mL

Chemical Identifiers

Chemical Name or Material Silicon, AAS standard solution
Name Note Si 1000μg/mL
Purity Grade Specpure™
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47277AE

  SDS
alfa aesar™
47277AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Silicon, AAS standard solution
Quantity 100mL
TSCA TSCA
Name Note Si 1000μg/mL
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00085311