Please login to your online account to display your discounted pricing

Silicon, +99%, powder, -325 mesh, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC201565000

 View more versions of this product

Catalog No. AC201565000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Fine Powder
Formula Weight 28.09
Quantity 500g
Color Dark gray
Merck Index 15, 8630
Chemical Name or Material Silicon
Name Note -325 mesh
Molecular Formula Si
CAS 7440-21-3
Assay Percent Range 99+%
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Flammable solid.
MDL Number MFCD00085311
Packaging Plastic bottle
Total Trace Metal Impurities 1.0% max.
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material Silicon
Name Note -325 mesh
CAS 7440-21-3
Physical Form Fine Powder
Assay Percent Range 99+%
Molecular Formula Si
Formula Weight 28.09
MDL Number MFCD00085311
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.

SDS & Safety Documents