Please login to your online account to display your discounted pricing

Silicon, +99%, powder, -325 mesh, ACROS Organics™

$18.17 - $219.21

Chemical Identifiers

Chemical Name or Material Silicon
Name Note -325 mesh
CAS 7440-21-3
Physical Form Fine Powder
Assay Percent Range 99+%
Molecular Formula Si
Formula Weight 28.09
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC201565000

  SDS
thermo fisher scientific
AC201565000
Plastic bottle 500g Each for $219.21

AC201560100

  SDS
thermo fisher scientific
201560100
Glass Bottle 10g Each for $18.17

AC201561000

  SDS
thermo fisher scientific
201561000
Plastic bottle 100g Each for $99.11
Description & Specifications

Specifications

Physical Form Fine Powder
Formula Weight 28.09
Quantity 500g
Color Dark gray
Merck Index 15, 8630
Chemical Name or Material Silicon
Name Note -325 mesh
Molecular Formula Si
CAS 7440-21-3
Assay Percent Range 99+%
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Flammable solid.
MDL Number MFCD00085311
Packaging Plastic bottle
Total Trace Metal Impurities 1.0% max.
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.