Please login to your online account to display your discounted pricing

Silicagel orange, for drying purposes, non toxic grade, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC392032500

 View more versions of this product

Catalog No. AC392032500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Beads
Quantity 250g
Color Orange
Chemical Name or Material Silica gel orange
MDL Number MFCD00077645
Loss on Drying 2% max. (120°C, 2 h)
Additional Information Adsorption capacity: 20% (25°C min., 50% rH)
Packaging Glass bottle
CAS 1327-36-2

Specifications

Chemical Name or Material Silica gel orange
CAS 1327-36-2
MDL Number MFCD00077645