Please login to your online account to display your discounted pricing

Silicagel, functionalized, Tetrabutylammonium chloride, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Silica gel, functionalized, tetrabutylammonium chloride
Name Note Functionalized, Tetrabutylammonium chloride
CAS 7631-86-9
CAS na
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD06660074
Synonym Tetrabutylammonium chloride, solid supported on silicagel
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375630050

  SDS
thermo fisher scientific
AC375630050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375630010

  SDS
thermo fisher scientific
AC375630010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 60.08
Quantity 5g
Color Pale yellow
Merck Index 15, 8632
Synonym Tetrabutylammonium chloride, solid supported on silicagel
Loading 0.6mmol/g exact loading on the certificate of analysis
CAS na
Chemical Name or Material Silica gel, functionalized, tetrabutylammonium chloride
Name Note Functionalized, Tetrabutylammonium chloride
Molecular Formula O2Si
MDL Number MFCD06660074
CAS 7631-86-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

For solid support synthesis

Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.