Please login to your online account to display your discounted pricing

Silicagel, functionalized, Diphenylphosphinoethyl, ca. 0.7mmol/g, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC376470050

 View more versions of this product

Catalog No. AC376470050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 60.08
Physical Form Fine Crystalline Powder
Quantity 5g
Color White to light yellow
Particle Size 40 to 63 Micron
Merck Index 15, 8632
Synonym Diphenylphosphinoethyl substituted silicagel
Health Hazard 1 Warning
MDL Number MFCD03095922
Chemical Name or Material Silica gel, functionalized, diphenylphosphinoethyl
CAS 7631-86-9
Name Note Functionalized, Diphenylphosphinoethyl, ca. 0.7mmol/g
Molecular Formula O2Si
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Loading (molecular) ca. 0.7mmol/g
CAS 607-01-2
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

Specifications

Chemical Name or Material Silica gel, functionalized, diphenylphosphinoethyl
Name Note Functionalized, Diphenylphosphinoethyl, ca. 0.7mmol/g
CAS 7631-86-9
CAS 607-01-2
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD03095922
Synonym Diphenylphosphinoethyl substituted silicagel
Accessories

Warning

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

SDS & Safety Documents