Please login to your online account to display your discounted pricing

Silicagel, functionalized, (cyclohexylcarbodiimido)propyl, ACROS Organics


Quantity 5g

Manufacturer: thermo fisher scientific  AC371530050

 View more versions of this product

Catalog No. AC371530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 60.08
Quantity 5g
Chemical Name or Material Silicagel
Synonym Cyclohexylcarbodiimide, immobilized on silicagel
MDL Number MFCD03701774
CAS 7631-86-9
Name Note Functionalized, (cyclohexylcarbodiimido)propyl, ca. 0.9mmol/g, particle size: 40-63 micron
Molecular Formula O2Si

Specifications

Chemical Name or Material Silicagel
Name Note Functionalized, (cyclohexylcarbodiimido)propyl, ca. 0.9mmol/g, particle size: 40-63 micron
CAS 7631-86-9
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD03701774
Synonym Cyclohexylcarbodiimide, immobilized on silicagel