Please login to your online account to display your discounted pricing

Silicagel, functionalized, 4-Bromophenyl, ca. 1 mmol/g, part. size: 40-63 micron, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Silica gel, functionalized, 4-bromophenyl
Name Note Functionalized, 4-Bromophenyl, ca. 1 mmol/g
CAS 7631-86-9
CAS 108-86-1
Physical Form Crystalline Powder
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD03096006
Synonym 4-Bromophenyl substituted silicagel
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376480010

  SDS
thermo fisher scientific
AC376480010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC376480050

  SDS
thermo fisher scientific
AC376480050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.08
Physical Form Crystalline Powder
Quantity 1g
Color White to off-white
Particle Size 40 to 63 Micron
Merck Index 15, 8632
Packaging Glass bottle
MDL Number MFCD03096006
Chemical Name or Material Silica gel, functionalized, 4-bromophenyl
Molecular Formula O2Si
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Loading (molecular) ca. 1mmol/g
CAS 108-86-1
Synonym 4-Bromophenyl substituted silicagel
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.
Name Note Functionalized, 4-Bromophenyl, ca. 1 mmol/g
CAS 7631-86-9

For solid support synthesis

Safety and Handling

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Toxic if inhaled.