missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica GF-TLC, 60A, EcoChrom™, MP Biomedicals™

Manufacturer:  Mp Biomedicals 0204645

 View more versions of this product

Catalog No. ICN04645



Description

Description

Specifications

Specifications

Silica
7631-86-9
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08
2g/cm3 at 20°C
60Å
500g
GF-TLC
O2Si
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
O=[Si]=O
60.083
CHEBI:30563
Powder
500-600m2/g
White to Gray

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

EINECSNumber : 231-545-4

Recommended Storage : Store at Room Temperature.

SDS
Documents
Product Certifications