missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica GF-TLC, 60A, EcoChrom™, MP Biomedicals™

$182.60 - $4,362.47

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN04645
View Documents
Mp Biomedicals
0204645
500g
Each for $182.60
 
ICN04680
View Documents
Mp Biomedicals
0204680
25kg
Each for $4,362.47
 
Description

Description

Specifications

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
CHEBI:30563
O=[Si]=O

Specifications

7631-86-9
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08
2g/cm3 at 20°C
60Å
500g
O2Si
aerosil, cristobalite, diatomaceous earth, infusorial earth, quartz, silica, silica, amorphous, silicic anhydride, silicon dioxide, tridymite
O=[Si]=O
60.083
CHEBI:30563
Powder
500-600m2/g
White to Gray
Silica, GF-TLC
Safety and Handling

Safety and Handling

EINECSNumber : 231-545-4

Recommended Storage : Store at Room Temperature.

SDS