missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silica gel, spherical, 300 angstroms, Thermo Scientific Chemicals
$289.75 - $954.48
Chemical Identifiers
CAS | 63231-67-4 |
---|---|
Molecular Formula | H2O3Si |
Molecular Weight (g/mol) | 78.098 |
MDL Number | MFCD00011232 |
InChI Key | IJKVHSBPTUYDLN-UHFFFAOYSA-N |
Synonym | metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid |
PubChem CID | 14768 |
IUPAC Name | dihydroxy(oxo)silane |
SMILES | O[Si](=O)O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
63231-67-4 | |
78.098 | |
IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
14768 | |
O[Si](=O)O |
H2O3Si | |
MFCD00011232 | |
metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid | |
dihydroxy(oxo)silane |
Specifications
63231-67-4 | |
H2O3Si | |
metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid | |
O[Si](=O)O | |
78.098 | |
Plastic bottle |
10 g | |
MFCD00011232 | |
IJKVHSBPTUYDLN-UHFFFAOYSA-N | |
dihydroxy(oxo)silane | |
14768 | |
Silica gel |
Safety and Handling
EINECSNumber : 231-545-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only