missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica gel, spherical, 100A, Alfa Aesar™

Manufacturer:  Alfa Aesar 4409309

 View more versions of this product

Catalog No. AA4409309



Specifications

Specifications

Silica gel
10g
H2O3Si
metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
O[Si](=O)O
78.098
Plastic bottle
63231-67-4
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
14768

Chemical Identifiers

63231-67-4
78.098
IJKVHSBPTUYDLN-UHFFFAOYSA-N
14768
O[Si](=O)O
H2O3Si
MFCD00011232
metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
dihydroxy(oxo)silane
Safety and Handling

Safety and Handling

TSCA : Yes

SDS Documents