missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silica gel, spherical, 100 angstroms

Manufacturer:  Alfa Aesar™ 4409109

 View more versions of this product

Catalog No. AA4409109



Specifications

Specifications

Silica gel
63231-67-4
H2O3Si
5 Micron APS, 100Å
O[Si](=O)O
78.098
Spherical, 5 Micron APS,S.A. 350m
P.V. 0.9cc/g
10g
MFCD00011232
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
14768
350m2/g

Chemical Identifiers

63231-67-4
78.098
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
H2O3Si
MFCD00011232
14768
O[Si](=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 231-545-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents