Fisher Chemical™ Silica Gel Sorbent (70-230 Mesh, Grade 60)
Manufacturer:  Fisher Chemical™ FLS82625
 View more versions of this product
Catalog No. S826-25
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Specifications
Specifications
63 to 200 µm
Fiber Drum
Silica Gel
60
25 kg