Please login to your online account to display your discounted pricing
Silica Gel Sorbent (35-70 Mesh, Grade 60), Fisher Chemical
Manufacturer: fisher scientific company  FLS834500
 View more versions of this product
Catalog No. S834-500
  • / Each
  • / Case of 6 Each
Description & Specifications
Specifications
Grade 60
Quantity 500g
Material Silica Gel
Particle Size 200 to 500μm