Please login to your online account to display your discounted pricing

Silica Gel SilicAR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Silica Gel
Name Note Grade 62 (60-200Mesh)
CAS 1343-98-2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK6447250

 
avantor performance
644702
Glass 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK6512250

 
avantor performance
651202
Glass 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK6551250

 
avantor performance
655102
Glass 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK6551P212

 
avantor performance
655105
Poly Pail 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Silica Gel
Packaging Glass
Name Note Grade 62 (60-200Mesh)
CAS 1343-98-2
Quantity 250g