missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel SilicAR, Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance 655102

 View more versions of this product

Catalog No. MK6551250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Silica Gel
1343-98-2
H4O4Si
RMAQACBXLXPBSY-UHFFFAOYSA-N
silicic acid
14942
Grade 62 (60-200Mesh)
250g
Glass
O[Si](O)(O)O
96.113
CHEBI:26675

Chemical Identifiers

1343-98-2
96.113
14942
silicic acid
H4O4Si
RMAQACBXLXPBSY-UHFFFAOYSA-N
CHEBI:26675
O[Si](O)(O)O
SDS Documents