Please login to your online account to display your discounted pricing

Silica Gel SilicAR, Macron Fine Chemicals™


macron

Quantity 250g
Packaging Glass

Manufacturer: avantor performance  651202

 View more versions of this product

Catalog No. MK6512250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Glass
Quantity 250g
Chemical Name or Material Silica Gel
Name Note Grade 62 (60-200Mesh)
CAS 1343-98-2

Specifications

Chemical Name or Material Silica Gel
Name Note Grade 62 (60-200Mesh)
CAS 1343-98-2