Please login to your online account to display your discounted pricing

Silica Gel, Grade 47 (10-18 Mesh/Certified ACS), Fisher Chemical


Quantity 50kg
Packaging Fiber Drum

Manufacturer: fisher scientific company  S16150

 View more versions of this product

Catalog No. S16150

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Quantity 50kg
Mesh Size 10 to 18 Mesh
Physical Form Solid
Color Blue
Chemical Name or Material Silica Gel
Molecular Formula SiO2
MDL Number MFCD00011232
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
CAS 7631-86-9
Identification Pass Test
Packaging Fiber Drum
Suitability Pass Test
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Name Note Tel-Tale™ Desiccant, Indicating, 10-18 Mesh
CAS 7646-79-9

Specifications

Chemical Name or Material Silica Gel
Name Note Tel-Tale™ Desiccant, Indicating, 10-18 Mesh
CAS 7646-79-9
CAS 7631-86-9
Purity Grade Certified ACS Grade
Physical Form Solid
Molecular Formula SiO2
MDL Number MFCD00011232
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Possible cancer hazard May cause cancer based on animal data. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1

CAUTION!

SDS & Safety Documents