Please login to your online account to display your discounted pricing

Silica gel, functionalized, diethylenetriamine tetraacetate sodium salt, 0.5mmol/g, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC372230050

 View more versions of this product

Catalog No. AC372230050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 60.08
Physical Form Powder
Quantity 5g
Color Light beige
Merck Index 15, 8632
CAS na
Loading 0.5mmol/g, exact loading on the certificate of analysis
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula O2Si
CAS 7631-86-9
Chemical Name or Material Silica gel, functionalized, diethylenetriamine tetraacetate sodium salt
Packaging Glass bottle
Name Note Functionalized, Diethylenetriamine tetraacetate sodium salt, 0.5 mmol/g
MDL Number MFCD06660073
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Silica gel, functionalized, diethylenetriamine tetraacetate sodium salt
Name Note Functionalized, Diethylenetriamine tetraacetate sodium salt, 0.5 mmol/g
CAS 7631-86-9
CAS na
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD06660073
Accessories

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents