Please login to your online account to display your discounted pricing

Silica gel, functionalized, diethylenetriamine tetraacetate sodium salt, 0.5mmol/g, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Silica gel, functionalized, diethylenetriamine tetraacetate sodium salt
Name Note Functionalized, Diethylenetriamine tetraacetate sodium salt, 0.5 mmol/g
CAS 7631-86-9
CAS na
Molecular Formula O2Si
Formula Weight 60.08
MDL Number MFCD06660073
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC372230010

  SDS
thermo fisher scientific
AC372230010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC372230050

  SDS
thermo fisher scientific
AC372230050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 60.08
Quantity 1g
Color Light beige
Merck Index 15, 8632
Name Note Functionalized, Diethylenetriamine tetraacetate sodium salt, 0.5 mmol/g
Chemical Name or Material Silica gel, functionalized, diethylenetriamine tetraacetate sodium salt
Molecular Formula O2Si
MDL Number MFCD06660073
CAS 7631-86-9
Loading 0.5mmol/g, exact loading on the certificate of analysis
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS na
Health Hazard 3 GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle

For solid support synthesis

Safety and Handling

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.